Revco® Blood Bank Refrigerators, 1 to 8 °C, Thermo Scientific
Supplier: Thermo Scientific
Revco®
REB404A REB1204-A REB3004A REB3004-D
CA14002-936 CA55702-710 CA55702-962 CA55702-960
Revco® Blood Bank Refrigerators, 1 to 8 °C, Thermo Scientific
Refrigerators
Designed to meet AABB, ANRC, and FDA standards for storage of whole blood and blood components
Refrigerators are equipped with an advanced defrost sensor to manage the defrost cycle, minimizing trace amounts of frost buildup to maintain refrigeration coil efficiency for responsive and economical perfomance.
Positive, forced air circulation maintains temperature uniformity at all shelf levels. Directional airflow assures quick recovery after door openings. Self-closing, lockable doors. All models except the undercounter unit feature glass doors and a seven-day temperature recorder.
Note: Actual blood bag storage capacities will vary according to loading method, size of containers, and loading preference.
Refrigerators have a nominal capacity of 48 to 576 blood bags with volumes of 450 ml. Units feature the IntrLogic™ microprocessor control system, positive airflow systems, and industrial quality cabinet construction. Extra-strength refrigeration compressors are designed for industrial, clinical, and scientific use. Temperature is factory preset to 4 °C and is adjustable from 1 to 8 °C. Icon-based controls and displays include graphic references for universal operation. A key-operated, triple-position master switch controls power to all systems for extra safety and setpoint security. Audible and visual alarms warn of power failure and temperature deviation of 5.5 °C above or 1.5 °C below setpoint. A push button alarm test and battery backup assure system readiness and sensor continuity. An extra-long alarm probe lead allows manual testing if necessary.
Certifications: UL and C-UL listed. CE marked.