Would you like to log in again?
2700 N95 Particulate Respirator, Moldex®
Supplier: MOLDEX/METRIC INC.
2700N95 2701N95
56219-012 56219-014
2700 N95 Particulate Respirator, Moldex®
Disposable respirator provides a filter efficiency level of 95% or greater against particulate aerosols free of oil up to 10 times OSHA Permissible Exposure Limit (PEL).
Stretchable HandyStrap® with buckle allows mask to hang around neck when not in use. Exclusive Ventex® valve vents hot air out for reduced breathing resistance. Soft foam nose flange eliminates pressure points. Dura-Mesh® shell resists collapsing in heat and humidity and keeps filter media clean-looking longer. Soft lining increases worker comfort. Available in two sizes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty