Would you like to log in again?
N100 Particulate Respirators, Moldex®
Supplier: MOLDEX/METRIC INC.
2730N100 2731N100
NIOSH-certified to have a 99.97% or greater filter efficiency against oil-free particulate aerosols
  • Stretch-cloth HandyStrap® with buckle to hang mask around neck when not in use
  • Ventex® exhale valve lets hot air escape quickly for easier breathing and extended use
  • Soft foam full flange relieves pressure points
  • Dura-Mesh® shell resists collapsing in heat and humidity
  • Flame retardants added to shell to decrease flammability
  • Softspun® lining for increased comfort and durability
Suggested for use in environments where an OSHA substance-specific standard applies (lead, cadmium, arsenic, MDA) as well as pharmaceutical manufacturing, welding, cutting, and metal pouring.
ORDER
Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
Medium/Large 2730N100 56219-276 Case of 30 Retrieving
Pack of 5 Retrieving
Product and/or packaging is safer to human and environmental health Small Business Enterprise
Small 2731N100 56219-274 Case of 30 Retrieving
Pack of 5 Retrieving
Product and/or packaging is safer to human and environmental health Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Environmentally Preferable Attribute Product and/or packaging is safer to human and environmental health
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty