VWR® Critical Cover® PFL® Face Masks
Supplier: Avantor
VWR® Critical Cover® PFL®
9908
47732-680CS 316.13 USD
47732-680
VWR® Critical Cover® PFL® Face Masks
Masks Hygiene Masks
Masks are hypoallergenic and fluid-resistant.
  • Hypoallergenic
  • Comfortable breathing chamber
  • Custom fit to any size face
The Twist-Seal® design utilizes an encased malleable aluminum band under the chin to provide a custom fit for every face. Masks are secured with a latex-free band. Also available cleanroom packaged.
Color: blue.
Ordering information: Cleanroom packaged masks are supplied 50 masks per bag, 6 bags per case.