MaxQ™ Reciprocating and Dual-Action Reciprocating/Orbital Shakers, Thermo Scientific
Supplier: Thermo Scientific
SHKA2508 SHKA2506
80085-934EA 3666.72 USD
80085-934 80085-938
MaxQ™ Reciprocating and Dual-Action Reciprocating/Orbital Shakers, Thermo Scientific
The reciprocating shaker provides gentle to vigorous agitation with variable 10–250rpm speed control and is ideal for extractions, mixing blood samples, and silver staining of polyacrylamide gels
The dual-action shaker provides gentle to vigorous agitation with variable 40–400rpm speed control and is ideal for growing bacteria, dissolution studies, staining of electrophoresis gels, and extractions. This shaker offers both reciprocating linear motion with a fixed stroke length of 1.3cm (1/2"), and orbital motion in a 1.3cm (1/2") diameter orbit. A control lever changes the shaking motion while unit is stationary or in operation.
The timer on both shakers can be set for 1–60 minutes or for continuous shaking. Units operate in temperature ranges from 4 to 40°C (39 to 104°F) and noncondensing humidity conditions from 20–80%, enabling them to be used in general-purpose incubators, warm rooms, environmental chambers, and refrigerators. Shakers can be fitted with 28x33cm (11x13"); 46x46cm (18x18"); or 46x61cm (18x24") shaking platforms to accommodate a variety of labware.
This shaker offers reciprocating linear motion with an adjustable stroke length of 1.3–3.8cm (1/2–11/2") to mimic hand shaking of separatory funnels.
Certifications: UL listed. The 240V dual-action shaker is CE marked.
Ordering information: Manufacturer's warranty is 1 year labor, 5 years parts, and lifetime on the drive mechanism. All shakers include bearing pans; platforms and clamps are ordered separately. For universal platforms, see the 57018-990 series; for dedicated platforms, see the 72890-180 and 72890-138 series; for clamps, see the 57019-670 series.