SureGrip™ Hook Clips, Model AC280, Fluke®
Supplier: Fluke
SureGrip™
1610115
26984-308EA 92.85 USD
26984-308
SureGrip™ Hook Clips, Model AC280, Fluke®
Electrical Sockets
Sold as a pair containing one red and one black clip
Clip profile narrows to 5.6 mm (7/32") at tip. Hook opening is 6.4 mm (1/4") at front. CAT III rated for 1000 V; CAT IV rated for 600 V, 3 A.