Metal Lockout Hasps, Honeywell Safety
Supplier: Honeywell Safety
R60ML MS86
11000-688EA 29.76 USD
CA11000-686 11000-688
Metal Lockout Hasps, Honeywell Safety
Security Locks
Metal lockout hasps are scissor type and constructed of tempered steel
Each hasp will accept six padlocks with up to 9.5mm (3/8in.) diameter shackles. Hasp 11000-688 has a rubber dipped handle. Hasps are painted red.