Would you like to log in again?
Spectra/Gel® Absorbent, Spectrum® Laboratories
Supplier: SPECTRUM LABORATORIES INC
Spectra/Gel®
292600
Designed to facilitate the concentration of protein sample solutions quickly and efficiently, this polyacrylate-polyalcohol absorbent expands as it absorbs water
After dialysis, simply remove the tubing containing the sample and coat it with Spectra/Gel® absorbent. Water and smaller molecules diffuse through the membrane and are absorbed. Concentration continues unattended until a predetermined volume of water is removed from the sample. The solution is easy to remove after absorbancy. The time needed for concentration will depend upon the final volume, the MWCO, the surface-to-volume ratio of the membrane tubing, and the amount of Spectra/Gel® absorbent solution applied.
The high molecular weight polymer can be used with dialysis membranes ranging from MWCO 100 to 300,000 without contaminating the sample.
ORDER
Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
500 g 292600 28170-552 Each Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty