Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Max System™ Rack, Rubbermaid®

Supplier: Janitorial Supplies
Max System™
RCP3320BLA
Large, black, end loader rack is ideal for a wide range of food preparation, transport, serving, and storage tasks

Features 18 slots for full size sheet pans and food boxes. Structural web polypropylene construction is impact-resistant and provides quiet handling. Rounded corners are soft for minimal impact on walls, doors, or other equipment. Wheels have two locking and two non-locking, non-marking swivel plate casters. Accommodates one 45.7x66cm (18x26") or two 45.7x33cm (18x13") pans per slot.

Certifications: USDA Meat and Poultry Group compliant. NSF Std. 2 certified.



 

ORDER

Expand All / Collapse All
Capacity Supplier No. VWR Catalog Number Unit Price Quantity
175 lbs. RCP3320BLA 89073-266 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Rack Dimensions 283/4" x 233/4" x 677/8"
Caster Size 5"

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping