HA Ultrogel™ Chromatography Resins, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
24775-082 24775-025 24775-025
82030-570EA 176.23 USD
82030-570 CA87004-866 87004-866
HA Ultrogel™ Chromatography Resins, Cytiva (Formerly Pall Lab)
These composite resins are composed of cross-linked agarose beads with micro-crystals of hydroxyapatite entrapped in agarose mesh.
  • Composite resins
  • Effective purification
  • Biomolecule selectivity
HA Ultrogel™ resins are an effective purification mechanism in a variety of processes, providing biomolecule selectivity complementary to traditional ion exchange or hydrophobic interaction techniques. Main applications are fractionation and purification of biomolecules from research and development scale to manufacturing. Ready-to-use, suspended in 1M sodium chloride with 20% ethanol as bacteriostatic. Capacity for cytochrome C determined using 5mg/mL cytochrome C diluted 50/50 in 10mM sodium phosphate buffer, pH 6.8 at 30cm/h. Capacity for BSA determined using 1mg/mL BSA diluted 50/50 in 10mM sodium phosphate buffer, pH 6.8 at 12.5cm/h.