Nalgene® Autoclavable Dolly, Stainless Steel, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2624-0020
16210-082CS 1024.42 USD
16210-082
Nalgene® Autoclavable Dolly, Stainless Steel, Thermo Scientific
Carts
Designed to move small NALGENE tanks (up to 115L/30gal.) and carboys during daily use or servicing
Resistant to chemical corrosion and rust. Accepts vessels up to 52.1cm (201/2") in diameter. Casters won't leave marks on floor.