VWR® Microplate Stability Chamber
930205
12621-182EA 1036.48 USD
12621-182
VWR® Microplate Stability Chamber
When placed inside a standard or CO2 incubator, this stability chamber works to reduce the variations often experienced in cell-based assays.
  • Now with Best-in-Class, Exclusive 5-Year Warranty
The chamber uses the incubator heat and atmosphere to create a homogeneous microenvironment for microwell plates rather than the heterogeneous environments created in traditional incubators. High humidity within the chamber eliminates evaporation from the plates that causes the edge effect. The thick anodized aluminum shelf distributes heat evenly to up to six plates for excellent temperature uniformity. The chamber includes a fan powered by a 120V power converter.
Ordering information: Additional aluminum microplate trays sold separately.