BigEasy® TSA™ Electrocompetent Cells, Biosearch Technologies
Supplier: Biosearch Technologies
Big Easy® TSA™
60224-1
95024-142EA 297.81 USD
95024-142
BigEasy® TSA™ Electrocompetent Cells, Biosearch Technologies
Competent Cells
Designed for use with the BigEasy® v2.0 linear cloning kit, the BigEasy® TSA™ electrocompetent cells provide high transformation efficiency and induction of copy number
Electrocompetent cells require immediate storage at –80°C (–112°F). The control and recovery medium can both be stored at –20°C (–4°F) or –80°C (–112°F).
This strain, derived from Lucigen's E.cloni® 10G strain, give high yield and high quality plasmid DNA due to the endA1 and recA1 mutations. They contain the mcr and mrr mutations, which allow methylated genomic DNA that has been isolated directly from mammalian or plant cells to be cloned without deletions or rearrangements. BigEasy® TSA™ cells are also resistant to infection by phage T1. Cells have an ampicillin gene integrated into the chromosome and no exogenous plasmids.
Ordering information: BigEasy® TSA™ electrocompetent cells are shipped on dry ice in one container along with supercoiled control pKanR DNA at 1ng/µL and recovery medium. For additional information on the BigEasy® v2.0 linear cloning kit, see 95024-148 series.