Inlet Sleeves for Varian GCs, Restek
20848 20898
RK20898PK 368.26 USD
RK20898 RK20848
Inlet Sleeves for Varian GCs, Restek
All sleeves are deactivated borosilicate glass.
Cyclosplitter® sleeve is a cylindrical glass screw design that provides a large area for sample vaporization and is ideal for dirty samples
Cyclosplitter® sleeve is easy to clean and many injections can be made before cleaning is required. Not recommended for large volume injections. Cup splitter sleeve features a Jenning’s cup design excellent for mixing of the sample. Tortuous flow path aids in sample vaporization and minimizes molecular weight discrimination. Can be packed with wool.
Splitless sleeve is for use with trace samples and for samples containing a narrow molecular weight distribution not prone to thermal decomposition. Packing with wool is recommended and aids in vaporization of high surface area for rapid vaporization of the sample delivering a uniform vapor cloud to the split point.
Frit splitter sleeve is for use with dirty, non-active samples.
Double gooseneck splitless sleeve is designed for better containment of the sample vaporization cloud and reduces sample backflash. Adsorption and discrimination are minimized. Can be packed with wool for dirty samples. For trace active samples.
Cyclo double gooseneck sleeve is ideal for trace active dirty samples.
Uniliner® sleeve is for use on trace, active samples requiring high recovery and linearity. Use the open-top for trace, dirty, active samples.
SPI sleeve is for 0.25 and 0.32 mm I.D. columns. Use when high linearity is desired.