KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
37033-10 37033-11100 37033-5
89002-702CS 302.56 USD
89002-702 89002-698 89002-700
KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Pipettes Serological Pipettes
Intended for bacteriological, microbiological, or serological work, these pipettes are graduated and calibrated to deliver to the tip.
They must be blown out to obtain total rated capacity. KIMAX N-51A® glass provides the best resistance to chemical attack and scratching. Scale is permanent brown stain fused into the surface of uniform bore tubing without etching. Pipettes are color-coded to assist in sorting and size selection. The 1 ml size has a plain fire-polished top, while the 5 and 10 ml sizes are tooled for cotton plugging. Designed from ASTM Specification E1044, Style I requirements.