You Searched For: Coomassie® Brilliant Blue R-250
12  results were found
SearchResultCount:"12"
Sort Results
List View Easy View
Rate These Search Results
Supplier: VWR
Description: Commonly used stain for the detection of protein bands following electrophoresis.
Catalog Number: (TCB3194-5G)
Supplier: TCI America
Description: CAS Number: 6104-59-2 MDL Number: MFCD00041762 Molecular Formula: C45H44N3NaO7S2 Molecular Weight: 825.97 Form: Crystal Color: Deep Red
Supplier: MP Biomedicals
Description: Coomassie Blue R-250 is a triphenylmethane dye used extensively in SDS-PAGE for the analysis of proteins.
Supplier: MP Biomedicals
Description: Coomassie® Brilliant Blue R-250 is a fast acting, sensitive dye, which can be used on SDS gels, IEF gels, and standard gels.
SDS
Catalog Number: (TCC0700-025G)
Supplier: TCI America
Description: CAS Number: 6104-59-2 MDL Number: MFCD00041762 Molecular Formula: C45H45N3O7S2 Molecular Weight: 825.97 Form: Crystal Color: Purple
Catalog Number: (EM-3340)
Supplier: MilliporeSigma
Description: Powder. Use for post-electrophoresis staining of proteins in applications requiring a high degree of sensitivity and/or solubility.
Supplier: Thermo Scientific Chemicals
Description: BRILLIANT BLUE R 100G
Catalog Number: (AAJ64297-18)
Supplier: Thermo Scientific Chemicals
Description: Crystalline powder
Catalog Number: (AAJ61384-K2)
Supplier: Thermo Scientific Chemicals
Description: Liquid
Catalog Number: (95043-426)
Supplier: G-Biosciences
Description: G-Biosciences' Coomassie Brilliant Blue (CBB) G-250 and R-250 Stain are based on a colloidal Coomassie stain. CBB G-250 and R-250 stain proteins with ...
SDS
Catalog Number: (95043-420)
Supplier: G-Biosciences
Description: G-Biosciences' Coomassie Brilliant Blue (CBB) G-250 and R-250 Stain are based on a colloidal Coomassie stain. CBB G-250 and R-250 stain proteins with ...
SDS
Catalog Number: (PI20278)
Supplier: Thermo Scientific
Description: Thermo Scientific Pierce Coomassie Brilliant Blue R-250 is one of the most common forms of coomassie dye, which is a key component of various colorime...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 12 of 12
no targeter for Bottom