You Searched For: Light Green SF Yellowish


13  results were found

SearchResultCount:"13"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (97061-558)
Supplier: VWR
Description: A valuable plasma stain used in contrast to hematoxylin. Also a counterstain to safranin.

Catalog Number: (EM1.15941.0100)
Supplier: MilliporeSigma
Description: Cas Number: 5141-20-8, 100G

SDS


Catalog Number: (TS61185-0250)
Supplier: Thermo Scientific Chemicals
Description: LIGHT GREEN SF YELLOWISH CERTIFIED 25G


Catalog Number: (TCL0141-001G)
Supplier: TCI America
Description: [as an indicator for assay of Food Yellow No.4 (Tartrazine)] CAS Number: 5141-20-8 MDL Number: MFCD00012121 Molecular Formula: C37H36N2O9S3 Molecu...

Supplier: Thermo Scientific Chemicals
Description: Useful as a general plasma stain, as a histological stain, and a cytological counterstain
Catalog Number: (TCL0054-025G)
Supplier: TCI America
Description: CAS Number: 5141-20-8 MDL Number: MFCD00012121 Molecular Formula: C37H36N2O9S3 Molecular Weight: 792.84 Form: Crystal Color: Deep Red

SDS


Supplier: Spectrum Chemicals
Description: Light Green SF Yellowish is a green triarylmethane dye. It is used in histology for staining collagen. It appears as brownish-red to purple powder and...
Catalog Number: (75810-382)
Supplier: Leica Biosystems
Description: Component of the Leica Modified GMS Silver Kit.

SDS


Supplier: Thermo Scientific Chemicals
Description: LIGHT GREEN SF YELLOWISH 100G

Supplier: Electron Microscopy Sciences
Description: Light Green SF Yellowish is a high quality ready to use solution. The solution is available as 0.2% in 0.2% acetic acid for use in the GM Method, as a...

Minority or Woman-Owned Business Enterprise

Supplier: Ambeed
Description: Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulfonatobenzyl)amino)phenyl)(4-sulfonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulfonate, Purity: 97%, CAS Number: 5141-20-8, Red to Dark red to Dark purple powder to crystal, Storage: Inert atmosphere, Room Temperature, Size: 1G

New Product

Catalog Number: (100504-078)
Supplier: Electron Microscopy Sciences
Description: Grocott's method for fungi. Dahl's method as a bone and calcium stain. McManus' method for glycogen. Fraser-Lendrum method as a connective tissue stai...

Minority or Woman-Owned Business Enterprise


Catalog Number: (101410-984)
Supplier: Electron Microscopy Sciences
Description: 1% Aqueous

Minority or Woman-Owned Business Enterprise


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 13 of 13
no targeter for Bottom