You Searched For: Nitro blue tetrazolium chloride (NBT)


13  results were found

SearchResultCount:"13"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR
Description: NITROBLUE TETRAZOLIUM CHL1G
Supplier: Thermo Scientific Chemicals
Description: NITRO BLUE TETRAZOLIUM CHLORIDE 500MG

Supplier: MP Biomedicals
Description: p-Nitroblue Tetrazolium Chloride is a substrate for dehydrogenases and other oxidases and precipitating agent for fibrinogen.

SDS

Supplier: MP Biomedicals
Description: p-Nitroblue Tetrazolium Chloride is an NADPH-diaphorase substrate that competitively inhibits NOS (nitric oxide synthase) (IC50 = 3-4 μM). In combinat...

SDS

Supplier: TCI America
Description: [for Biochemical Research] CAS Number: 298-83-9 MDL Number: MFCD00012159 Molecular Formula: C40H30Cl2N10O6 Purity/Analysis Method: 98.0% (HPLC...
Catalog Number: (AAB23792-02)
Supplier: Thermo Scientific Chemicals
Description: 90%. 0.5g.

Supplier: MilliporeSigma
Description: NADPH-diaphorase substrate that competitively inhibits nitric oxide synthase (IC50=3–4µM). A well-known scavenger of superoxide anions. Useful as a su...
Supplier: Electron Microscopy Sciences
Description: A substrate for dehydrogenases and other oxidases.

Minority or Woman-Owned Business Enterprise

Supplier: Biotium
Description: is an oxidant widely used in conjuction with BCIP or BCIP derivatives to detect phosphotase activity or used as a substrate for dehydrogenase and other oxidases to assess cell viability, proliferation and cytotoxicity. NBT forms a dark blue-colored precipitate after enzymatic reduction.

Catalog Number: (BT140245-500MG)
Supplier: BeanTown Chemical
Description: Nitro Blue Tetrazolium chloride, Grade: Technical, Purity: 90%, CAS Number: 298-83-9, Molecular Formula: C40H30N10O6. 2Cl, Synonym: Nitrotetrazolium Blue chloride, Nitro BT, Size: 500MG

SDS


Supplier: Thermo Scientific Chemicals
Description: Intense blue dye used in histochemistry determinations
Catalog Number: (100270-436)
Supplier: Indofine Chemical Company
Description: Products for Life Science 298-83-9 100mg NBT 817.60 +4[degree]C.

Minority or Woman-Owned Business Enterprise Small Business Enterprise


Supplier: Adipogen
Description: NADPH-diaphorase substrate that competitively inhibits NOS (nitric oxide synthase). Well-known scavenger of superoxide anions. Dye that is used for de...

Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 13 of 13
no targeter for Bottom