    Sort by:
  • Search Within Results
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  MP Biomedicals
Description:   CHAPSO is the hydroxy analog of CHAPS; a non-denaturing zwitterionic detergent for solubilizing proteins, especially integral membrane proteins while ...