Lauda® Immersion Cooler, Model ETK 30, Brinkmann™
L001958 27-65-882-2
80078-566EA 7015.05 USD
80078-566 CA80078-566
Lauda® Immersion Cooler, Model ETK 30, Brinkmann™
Portable, compact immersion cooler with environmentally-safe, CFC-free refrigerant cools reservoirs up to 6L to –40°C
Ideal for rotary evaporator cold trap condensers, cooling open reservoirs, vapor and solvent trapping, and impact testing. For temperature control, the immersion cooler can be used in conjunction with an immersion heating circulator. Cooler also eliminates constant cold trap hassles and the need for chemical refrigerants such as dry ice and acetone mixtures.
Ordering information: Includes a 5.1dia.x12.4Lcm (2x429/32") stainless steel coil that easily adapts to most open reservoirs and a 1.5m (429/32') flexible hose.