Wiretrol® I Disposable Micropipets, Drummond
Supplier: Drummond
Wiretrol® I Wiretrol®
5-000-1020 5-000-1025 5-000-1001 5-000-1010 5-000-1005 5-000-1050 5-000-1100
53480-458EA 40.82 USD
53480-458 53480-505 53480-287 53480-403 53480-356 53480-550 53480-651
Wiretrol® I Disposable Micropipets, Drummond
Pipets Microcapillaries
The Drummond Wiretrol provides the accuracy and high speed action generally found inexpensive microdispensers at a fraction of the cost.
  • Economical Positive-Displacement Micropipet with Fingertip Control
  • Ideal for a Wide Variety of Applications
  • Stainless Steel Plunger
  • Glass Micropipets
  • Fast and Accurate
A unique combination of a disposable precision bore calibrated micropipet and a stainless steel plunger with a Fingertip handle, the Wiretrol accepts both high and low viscosity liquids including corrosives.
It's perfect for a wide range of uses.
Accessories information: Additional plungers are available separately.
Ordering information: This kit includes 100 glass micropipets and one reusable plunger.