BRAND Transferpette® S Single-Channel Pipettors, BrandTech
Supplier: BrandTech
704774 704770 704772 704780 704768 704778 704769
89042-180EA 482.31 USD
89042-180 89042-182 89239-634 89042-172 89042-174 89042-176 89042-178
BRAND Transferpette® S Single-Channel Pipettors, BrandTech
Transferpette® S pipettors deliver an exceptional level of comfort and performance that results from increased attention to ergonomic, operational, and material innovations.
  • Transferpette® S offers simple one-handed operation of all functions
  • Four-digit volume display is always visible
  • The 12.5 mm pipetting stroke requires the shortest range of motion available
  • Advanced materials are utilized to achieve a robust yet lightweight, corrosion-resistant design
An easily-adjustable volume-setting wheel is accessible without shifting hand position, while a convenient brake securely sets the selected volume. Pipettors also feature a slim, corrosion-resistant tip ejector, covered nameplate, and BRAND's Easy Calibration™ technology (adjustable volume models only) for lasting reliability. Models do not require tools for calibration or disassembly. Pipettors are CE-IVD compliant and fully autoclavable at 121 °C (250 °F).
These are ideal manual pipettors for demanding laboratory applications run by operators who prefer a center piston-button design.
Accessories information: The accessory shelf/rack mount allows pipettors to conveniently rest on a shelf or rack. The carousel stand provides a convenient holding area and ready access for up to six pipettors.
Ordering information: All single-channel pipettors are supplied with a manufacturer's three-year warranty, shelf/rack mount, tip sample, performance certificate, and operating manual. Starter kits include three pipettors, three shelf/rack mounts, and three boxes of appropriately sized tips.