C.O.D. Digestion Reagent Vials, WTW
Supplier: WTW
253 113 253 121 253 112 253 133 253 122 253 123 253 111 253 110 253 132
80094-560EA 496.62 USD
80094-560 80094-576 80094-554 80094-566 80094-574 80094-564 80094-558 80094-556 80094-568
C.O.D. Digestion Reagent Vials, WTW
Environmental Test Kits
Dichromate reactor digestion reagents for fast, easy, safe determination of low-, mid-, and high-range C.O.D
Reagents are available containing mercuric sulfate to eliminate chloride interferences or mercury-free. Reagents with mercuric sulfate are U.S. EPA approved for method 410.4 C.O.D. testing.
Premeasured reagent is packaged in flat-bottom ampules that are compatible with most spectrophotometers. Reagents have a two-year shelf life.
Certifications: A certificate is provided to indicate traceability to standards provided by the National Institute of Standards and Technology.