Aluminum Heat Dispersion Plates, Expedeon
CPA20-020 CPA20-100 CPA20-050 CPA33-080A CPA20-080A CPA20-030 CPA33-050 CPA33-040A CPA33-100 CPA20-040A CPA33-030 CPA20-060 CPA33-020 CPA33-060
CBCPA20-050EA 123.68 USD
CBCPA20-050 CBCPA33-080A CBCPA20-060 CBCPA20-030 CBCPA33-050 CBCPA20-040A CBCPA33-060 CBCPA33-030 CBCPA20-020 CBCPA20-100 CBCPA33-020 CBCPA33-100 CBCPA33-040A CBCPA20-080A
Aluminum Heat Dispersion Plates, Expedeon
For use with adjustable nucleic acid sequencers.
Designed to clamp behind the glass plate sandwich for even heat distribution during analysis. Plates are machined ultra-flat to a 0.64 cm (1/4") thickness and are electrically nonconductive.