Barnstead/Thermolyne Sterilizer Temperature Indicators, Thermo Scientific
Supplier: Thermo Scientific
Barnstead/Thermolyne
AY759X2
55710-020PK 121.84 USD
55710-020
Barnstead/Thermolyne Sterilizer Temperature Indicators, Thermo Scientific
Sterilization Indicators
Temperature verification strips verify that a sterilizer has reached the proper temperature through a color change
Chemical indicator strips 55710-020, for use with steam sterilizers, require a temperature of 132 to 135°C (270 to 275°F). Chemitest™ indicator strips 58939-984, for use with Chemiclave® sterilizers, require a temperature of 132 to 136°C (270 to 277°F). Daily verification ensures that items being processed have been run through a cycle, preventing the accidental use of items that have not been adequately processed.