VWR® Wide Orifice Pipette Tips
Supplier: VWR
VWR®
1042-960-306 1199-965-306
89049-168CS 1118.53 USD
89049-168 89049-164
VWR® Wide Orifice Pipette Tips
Pipette Tips
Wide orifice tips are designed with a significantly larger opening to allow the pipetting of viscous solutions such as milk, syrup, or rDNA suspensions.
  • Inert polypropylene construction
  • Wide-orifice aerosol filter tips
  • Slimmer, longer design fits most microcentrifuge tubes
  • Autoclavable
The larger opening prevents the shearing of macrophage cells. Tips feature a slimmer, longer design to fit into most 0.5 ml microcentrifuge tubes or 12×75 mm culture tubes. Tips are constructed of inert virgin CFR 21 compliant polypropylene, and will fit most 100 to 1250 µl pipettors. Aerosol filter wide orifice tips hold 1000 µl under filter.
Ordering information: Pak Rak® racked tips include 576 tips per refill. Bagged tips are supplied in resealable bags of 1000 tips. Hinged racks are supplied with 96 tips per rack, 6 racks per pack.