Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® and VWR Signature™ 10 µL Extended Length Pipet Tips

Supplier: VWR International
VWR® VWR Signature™
1162-290-320 1162-965-306 1162-960-306 1036-800-000 1036-965-306 1162-800-000 1036-960-306 1036-290-320
Low-binding tips feature a modified polypropylene resin that limits the adhesion of cells, rDNA, and other viscous materials.

  • For use with most micropipettors

Extended length tips allow you to reach the bottom of most 0.65 mL microtubes. The top mounting port features a solid ring that prevents the pipettor barrel from touching the sides of microtubes, reducing the potential for contamination. The tips have an unbeveled, ultrafine dispensing end to allow the dispensing of sample volumes below 1µL. All tips are RNase- and DNase-free. Low-binding tips are also free of protease and human DNA. Sterile tips are endotoxin-free. Tips and hinged racks are autoclavable.

Ordering information: Pak Rak® Express refills contain pipet tips to fill 8x12 racks, without the packaging waste of standard tip boxes. Pak Rak Express refills require the Pak Rak system loader (see 30128-374 series), which dispenses tips quickly and easily from the refill pack into each rack, eliminating risk of contamination from hands or gloves.



 

ORDER

Expand All / Collapse All
Description Volume Length Sterility Packed VWR Catalog Number Unit Price Quantity
VWR Signature Low-Binding Tips 0.1–10 µL 3.8 cm (1.500") Sterile 96/Hinged Rack 37001-166 Case of 9600 Retrieving
Pack of 960 Retrieving
VWR Signature Low-Binding Tips 0.1–10 µL 3.8 cm (1.500") Nonsterile 96/Hinged Rack 37001-164 Case of 9600 Retrieving
Pack of 960 Retrieving
VWR Signature Low-Binding Tips 0.1–10 µL 3.8 cm (1.500") Nonsterile Bulk 37001-170 Case of 10000 Retrieving
Pack of 1,000 Retrieving
VWR Signature Low-Binding Tips 0.1–10 µL 3.8 cm (1.500") Nonsterile Pak Rak Express Refill 37001-168 Case of 13440 Retrieving
Pack of 1,344 Retrieving
VWR Tips 0.1–10 µL 3.8 cm (1.500") Sterile 96/Hinged Rack 46620-318 Case of 9600 Retrieving
Pack of 960 Retrieving
VWR Tips 0.1–10 µL 3.8 cm (1.500") Nonsterile 96/Hinged Rack 46620-316 Case of 9600 Retrieving
Pack of 960 Retrieving
VWR Tips 0.1–10 µL 3.8 cm (1.500") Nonsterile Bulk 46620-312 Case of 10000 Retrieving
Pack of 1,000 Retrieving
VWR Tips 0.1–10 µL 3.8 cm (1.500") Nonsterile Bulk 46620-314 Case of 13440 Retrieving
Pack of 1,344 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is recyclable or reduces waste, Product and/or packaging is safer to human and environmental health, Product is manufactured with low CO₂ impact

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty