Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Bevel Point Pipet Tips in Stack Racks

Supplier: VWR International
VWR®
1093-995-336 1093-990-336 1093-700-326 1093-990-326 1093-700-316 1030-990-306 1030-700-326 1030-990-326 1030-700-316 1093-990-306 1032-700-018
Stack Racks provide a cost-effective and environment-conscious alternative to racked tips.

  • For single-channel and multichannel pipettors
  • Half the weight of conventional racks
  • Stack racks provide same amount of tips with less plastic

Stackable racks of tips in the 8x12 (96 tips) or 8x24 (192 tips) configuration utilize the same bottom and lift-off lid, minimizing unnecessary plastic. Lid and bottom are hinged for use as a storage box when empty, or may be reused with Stack Rack refills. Storage box can be autoclaved after tip use. Tips are DNase- and RNase-free. Sterile tips are endotoxin-free. Graduated tips are marked at 10, 20, 50, 100, and 200µL.



 

ORDER

Expand All / Collapse All
Description Volume Length Tip Color Sterility Packed VWR Catalog Number Unit Price Quantity
Graduated Tips 1–200 µL 5 cm (1.955") Clear Nonsterile Refill for 53509-010 53509-012 Case of 9600 Retrieving
Pack of 960 Retrieving
Graduated Tips 1–200 µL 5 cm (1.955") Clear Nonsterile 96/Rack, 16 Racks/Pack 30128-388 Case of 15360 Retrieving
Pack of 1,536 Retrieving
Graduated Tips 1–200 µL 5 cm (1.955") Clear Nonsterile 96/Rack, 10 Racks/Pack 53509-010 Case of 9600 Retrieving
Pack of 960 Retrieving
Graduated Tips 1–200 µL 5 cm (1.955") Clear Nonsterile 192/Rack, 5 Racks/Pack 53509-004 Case of 19200 Retrieving
Pack of 960 Retrieving
Graduated Tips 1–200 µL 5 cm (1.955") Clear Nonsterile Refill for 53509-004 53509-005 Case of 4800 Retrieving
Pack of 960 Retrieving
Plain Tips 1–200 µL 5 cm (1.955") Yellow Nonsterile 96/Rack, 10 Racks/Pack 53509-815 Case of 9600 Retrieving
Pack of 960 Retrieving
Plain Tips 1–200 µL 5 cm (1.955") Yellow Nonsterile 192/Rack, 5 Racks/Pack 53512-258 Case of 19200 Retrieving
Pack of 960 Retrieving
Plain Tips 1–200 µL 5 cm (1.955") Yellow Nonsterile Refill for 53509-815 53512-260 Case of 9600 Retrieving
Pack of 960 Retrieving
Plain Tips 1–200 µL 5 cm (1.955") Yellow Nonsterile Refill for 53512-258 53512-257 Case of 4800 Retrieving
Pack of 960 Retrieving
Plain Tips for Eppendorf® Pipettors 1–100 µL 5 cm (1.955") Yellow Nonsterile 192/Rack, 5 Racks/Pack 53512-315 Case of 9600 Retrieving
Pack of 960 Retrieving
Graduated Tips 1–200 µL 5 cm (1.955") Clear Sterile 96/Rack, 160 Racks/Case 37000-698 Case of 15,360 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is recyclable or reduces waste, Product is manufactured with low CO₂ impact

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty