These tips provide maximum sample recovery and minimal fluid retention.
- For use with most pipettors
- Sterile tips are endotoxin-free
- Tips and hinged racks are autoclavable
- All tips are free of RNase, DNase, and trace metals
Transparent polypropylene permits precise viewing of samples. Longer, thinner tip with ultrafine point helps eliminate sample hang-up and allows access to small tubes and deep well plates. The outer wall of the graduated tips has flush calibration lines that eliminate hang-up on specimen tubes. The graduated tips also have markings at 250, 500, 750, and 1000 µl.
Ordering information: Stack Rack refills are stackable racks of tips that use the same bottom and lift-off lid. Stack Racks provide a cost-effective and environment-conscious alternative to racked tips, minimizing unnecessary plastic.