Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BRAND Transferpette® S Multichannel Pipettors, BrandTech

Supplier: BRANDTECH SCIENTIFIC INC.
Transferpette®
703708 703728 703706 703726 703712 703710 703732 703700 703730 703720
These multichannel pipettors have been benchmarked against leading industry instruments to provide a comfortable, easy-to-operate device with excellent accuracy and reproducibility. The Transferpette® S series combines advanced ergonomic design and innovative materials to create a durable, ultra lightweight, and high-performance pipettor.

One-handed volume adjustments can be easily made by thumb control without shifting hand position. Settings can be securely locked in place. The short pipetting stroke length of 12.5mm is designed to reduce instances of thumb hyperextension. The manifold rotates in either direction to allow users to optimize their hand position. A finger rest reduces static strain and a covered nameplate offers simple and quick instrument identification. Tip cone V-rings reduce tip mounting/ejecting forces and enhance tip compatibility. A four-digit display is always visible. Calibration time is significantly reduced with BRAND's Easy Calibration™ technology, which requires no tools. Individually removeable and replaceable tip cones and seals allow easy, in-lab cleaning and servicing without recalibration. Pipettors are completely autoclavable at 121°C (250°F) for 20 minutes.

Ordering information: Each instrument is supplied with a shelf/rack mount, a box of appropriate pipettor tips with two refills, a set of replacement V-rings, a performance certificate, and operating manual.



 

ORDER

Expand All / Collapse All
Description Volume Accuracy Precision Supplier No. VWR Catalog Number Unit Price Quantity
12-Channel Pipettor 10–100 µL ±0.8% ≤0.3% 703728 89080-140 Each Retrieving
12-Channel Pipettor 20–200 µL ±0.8% ≤0.3% 703730 89080-142 Each Retrieving
12-Channel Pipettor 30–300 µL ±0.6% ≤0.3% 703732 89080-144 Each Retrieving
12-Channel Pipettor 5–50 µL ±0.8% ≤0.4% 703726 89080-138 Each Retrieving
12-Channel Pipettor 0.5–10 µL ±1.6% ≤1.0% 703720 89080-136 Each Retrieving
8-Channel Pipettor 0.5–10 µL ±1.6% ≤1.0% 703700 89080-126 Each Retrieving
8-Channel Pipettor 5–50 µL ±0.8% ≤0.4% 703706 89080-128 Each Retrieving
8-Channel Pipettor 10–100 µL ±0.8% ≤0.3% 703708 89080-130 Each Retrieving
8-Channel Pipettor 20–200 µL ±0.8% ≤0.3% 703710 89080-132 Each Retrieving
8-Channel Pipettor 30–300 µL ±0.6% ≤0.3% 703712 89080-134 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Tip Cones with V Rings for Transferpette® -8/-12, 2-20 µL 705678 89400-218 Pack of 2 Retrieving
Tip Cones with V Rings for Transferpette® -8/-12, 2.5-25 µL 703333 89400-176 Pack of 2 Retrieving
Tip Cones with V Rings for Transferpette® -8/-12 and S-8/-12, 0.5-10 µL 705677 89400-216 Pack of 2 Retrieving
Filter 704652 89370-104 Pack of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty