Nunc® Open Well Upper Structures, Thermo Scientific
256665 256665
82029-738CS 543.47 USD
82029-738 CA82029-738
Nunc® Open Well Upper Structures, Thermo Scientific
For 96--well plates
Used for bonding to specialty base material, holding PCR plates, or adding agar to individual wells. Upper structure is constructed of polystyrene.