Would you like to log in again?
Pierce™ Heterobifunctional Cross Linkers, Thermo Scientific
Supplier: Thermo Scientific
Pierce
22317 22111 22112 22311 22113 22312 22114 22327 22329 21111 22363 23013 22122 33043 22322 22324 21558 28100 22360 77149 22362 22416 22339 22615 22297 22298 22299 22610 26099 22295 22305 22107 22349 22108 22306 22307 22308 21651 22309 21650 21451 22980 22981 22101 22102 33063 21857 22301 22103 22104 22622 22105 22589 22106 26100 26102
Cross linkers are used for the identification of near-neighbor protein relationships and ligand-receptor interactions.
Heterobifunctional cross linkers have two different reactive ends and are used in two step sequential reactions, where the least labile reactive end is reacted first. Heterobifunctional agents allow more controlled two step reactions, which minimizes undesirable intramolecular cross-reaction and polymerization. The most widely used heterobifunctional cross linking agents are used to couple proteins through amine and sulfhydryl groups.
ORDER
Expand All / Collapse All
Description Size Molecular Weight Reactive Group Spacer Arm Target Group Supplier No. VWR Catalog Number Unit Price Quantity
AEDP (3-[(2-Aminoethyl)dithio]propionic Acid Hydrochloride) 50 mg 217.74 Amine, Carboxyl 9.5 Å Amines/Hydrazides, NHS Esters/Sulfo-NHS Esters 22101 PI22101 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
AMAS, N-(α-Maleimidoacetoxy)-Succinimide Ester 50 mg 252.18 Maleimide, NHS Ester 4.4 Å Amines, Sulfhydryls 22295 PI22295 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
ANB-NOS, N-5-Azido-2-nitrobenzyloxy-succinimide 50 mg 305.20 NHS Ester, Nitrophenyl Azide 7.7 Å Amines, Nonspecific Sites 21451 PI21451 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
BMPH, N-(β-Maleimidopropionic acid)Hydrazide 50 mg 297.19 Hydrazide, Maleimide 8.1 Å Carbonyl Aldehydes, Sulfhydryls 22297 PI22297 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
BMPS, N-(β-Maleimidopropyloxy)Succinimide Ester 50 mg 266.21 Maleimide, NHS Ester 6.9 Å Amines, Sulfhydryls 22298 PI22298 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
EDC (1-ethyl-3-(3-dimethylaminopropyl)Carbodiimide Hydrochloride) 5 g 191.70 Carboxyl 0 Å Amine, Carboxyl 22980 PI22980 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
EDC (1-ethyl-3-(3-dimethylaminopropyl)Carbodiimide Hydrochloride) 25 g 191.70 Carboxyl 0 Å Amine, Carboxyl 22981 PI22981 Each (25g) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
EDC (1-ethyl-3-(3-dimethylaminopropyl)Carbodiimide Hydrochloride) 10 mg 191.70 Carboxyl 0 Å Amine, Carboxyl 77149 PI77149 Each (10mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
EMCA (N-Ɛ-Maleimidocaproic Acid) 1 g 211.21 Maleimide, Carboxyl 9.4 Å Amines, Sulfhydryls 22306 PI22306 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
EMCH, N-(Ɛ-Maleimidocaproic acid)Hydrazide 50 mg 225.24 Maleimide, Hydrazide 11.8 Å Carbonyls, Sulfhydryls 22106 PI22106 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
EMCS (N-(Ɛ-Maleimidocaproyloxy)Succinimide Ester) 50 mg 308.29 Maleimide, NHS Ester 9.4 Å Amines, Sulfhydryls 22308 PI22308 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
GMBS, N-(γ-Maleimidobutyryloxy)Succinimide Ester 50 mg 280.23 Maleimide, NHS Ester 10.2 Å Amines, Sulfhydryls 22309 PI22309 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
KMUH, N-(ĸ-Maleimidoundecanoic acid)Hydrazide 50 mg 295.38 Hydrazide, Maleimide 19 Å Carbonyls, Sulfhydryls 22111 PI22111 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
LC-SMCC (Succinimidyl-4-N-maleimidomethyl- cyclohexane-1-carboxy(6-amidocaproate)) 50 mg 447.48 Maleimide, NHS Ester 16.1 Å Amines, Sulfhydryls 22362 PI22362 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
LC-SPDP (Succinimidyl-6[3'-(2-pyridyldithio)-propion amido]Hexanoate) 50 mg 425.52 NHS Ester, Pyridyldisulfide 15.7 Å Amines, Sulfhydryls 21651 PI21651 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
L-Photo-Leucine 100 mg 143.15 Diazirine 22610 PI22610 Each (100mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
L-Photo-Methionine 100 mg 157.17 Diazirine 22615 PI22615 Each (100mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
MBS (m-Maleimidobenzoyl-N-hydroxysuccinimide Ester) 50 mg 314.25 Maleimide, NHS Ester 9.9 Å Amines, Sulfhydryls 22311 PI22311 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
MPBH (4-(4-Maleimidophenyl)butyric Acid Hydrazide Hydrochloride) 50 mg 309.75 Hydrazide, Maleimide 17.9 Å Carbohydrates, Sulfhydryls 22305 PI22305 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
View MSDS SDS
PDPH (3-(2-Pyridyldithio)propionyl Hydrazide) 50 mg 229.32 Hydrazide, Pyridyldisulfide 9.2 Å Carbohydrates, Sulfhydryls 22301 PI22301 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
PMPI (N-(p-Maleimidophenyl)Isocyanate) 50 mg 214.18 Isocyanate, Maleimide 8.7 Å Hydroxyls, Sulfhydryls 28100 PI28100 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SAT(PEO)4 (S-Acetyl-PEO4-NHS) 100 mg 421.46 NHS Ester and Thioacetyl-Protected Sulfhydryl 18.25 Å Amines, Maleimide/ Iodoacetyl, Vinyl Sulfone Cross-Linkers 26099 PI26099 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SATA (N-Succinimidyl S-Acetylthioacetate) 50 mg 231.23 NHS Ester, Thioacetyl-Protected Sulfhydryl 2.8 Å Amines, Maleimide/ Iodoacetyl, Vinyl Sulfone Cross-Linkers 26102 PI26102 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SATP (N-Succinimidyl S-Acetylthiopropionate) 50 mg 245.25 NHS Ester and Thioacetyl-Protected Sulfhydryl 4.1 Å Amines, Maleimide/ Iodoacetyl, Vinyl Sulfone Cross-Linkers 26100 PI26100 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SBAP (Succinimidyl 3-(Bromoacetamido)Propionate) 50 mg 307.10 Bromoacetyl and NHS Ester 6.2 Å Amines, Sulfhydryls 22339 PI22339 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SIA (N-Succinimidyl Iodoacetate) 50 mg 283.02 Iodoacetyl and NHS Ester 1.5 Å Amines, Sulfhydryls 22349 PI22349 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SIAB (N-Succinimidyl(4-iodoacetyl)Aminobenzoate) 50 mg 402.14 Iodoacetyl and NHS Ester 10.6 Å Amines, Sulfhydryls 22329 PI22329 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
View MSDS SDS
SM(PEG)6, NHS-PEG6-Maleimide 100 mg 601.60 Maleimide, NHS Ester 32.5 Å Amines, Sulfhydryls 22105 PI22105 Each (100mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SM[PEG]12, NHS-PEG12-Maliemide 1 g 865.92 Maleimide, NHS Ester 53.4 Å Amines, Sulfhydryls 22113 PI22113 Each (1g) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SM[PEG]12, NHS-PEG12-Maliemide 100 mg 865.92 Maleimide, NHS Ester 53.4 Å Amines, Sulfhydryls 22112 PI22112 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SM[PEG]2, NHS-PEG2-Maliemide 100 mg 425.39 Maleimide, NHS Ester 17.6 Å Amines, Sulfhydryls 22102 PI22102 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SM[PEG]2, NHS-PEG2-Maliemide 1 g 425.39 Maleimide, NHS Ester 17.6 Å Amines, Sulfhydryls 22103 PI22103 Each (1g) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SM[PEG]24, NHS-PEG24-Maliemide 100 mg 1394.55 Maleimide, NHS Ester 95.2 Å Amines, Sulfhydryls 22114 PI22114 Each (100mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SM[PEG]4, NHS-PEG4-Maliemide 100 mg 425.39 Maleimide, NHS Ester 17.6 Å Amines, Sulfhydryls 22104 PI22104 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SM[PEG]4, NHS-PEG4-Maliemide 1 g 513.50 Maleimide, NHS Ester 24.6 Å Amines, Sulfhydryls 22107 PI22107 Each (1g) Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
View MSDS SDS
SM[PEG]8, NHS-PEG8-Maliemide 100 mg 389.71 Maleimide, NHS Ester 39.2 Å Amines, Sulfhydryls 22108 PI22108 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SMCC (succinimidyl 4-(N-maleimidomethyl)cyclohexane-1-carboxylate) 50 mg 334.32 Maleimide and Sulfo-NHS Ester 8.3 Å Amines, Sulfhydryls 22360 PI22360 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SMPB (Succinimidyl 4-(p-Maleimidophenyl)Butyrate) 50 mg 356.33 Maleimide and NHS Ester 11.6 Å Amines, Sulfhydryls 22416 PI22416 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SMPH (Succinimidyl-6-([beta]-maleimidopropionamido)Hexanoate) 50 mg 379.36 Maleimide and NHS Ester 14.3 Å Amines, Sulfhydryls 22363 PI22363 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SMPT, 4-Succinimidyloxycarbonyl-methyl-α-(2-pyridyldithio)Toluene 50 mg 388.46 NHS Ester and Pyridyldisulfide 20.0 Å Amines, Sulfhydryls 21558 PI21558 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SPB (Succinimidyl-[4-(psoralen-8-yloxy)]Butyrate) 50 mg 385.32 NHS Ester and Psoralen 8.6 – 9.5 Å Amines, Thymine 23013 PI23013 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
SPDP (N-Succinimidyl 3-(2-Pyridyldithio)Propionate) 50 mg 312.37 NHS Ester and Pyridyldisulfide 6.8 Å Amines, Sulfhydryls 21857 PI21857 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Sulfo-EMCS (N-(Ɛ-Maleimidocaproyloxy)Sulfosuccinimide Ester) 50 mg 410.33 Maleimide and Sulfo-NHS Ester 9.4 Å Amines, Sulfhydryls 22307 PI22307 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Sulfo-GMBS, N-(γ-Maleimidobutryloxy)Sulfosuccinimide Ester 50 mg 382.28 Maleimide, Sulfo-NHS Ester 6.8 Å Amines, Sulfhydryls 22324 PI22324 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Sulfo-KMUS, N-(ĸ-Maleimidoundecanoyloxy)Sulfosuccinimide Ester 50 mg 480.47 Maleimide, Sulfo-NHS Ester 15.7 Å Amines, Sulfhydryls 21111 PI21111 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Sulfo-LC-SPDP (Sulfosuccinimidyl 6-[3'-(2-pyridyldithio)- propionamido]Hexanoate) 50 mg 527.57 Pyridyldisulfide, Sulfo-NHS Ester 15.6 Å Amines, Sulfhydryls 21650 PI21650 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Sulfo-MBS (m-Maleimidobenzoyl-N-hydroxysulfosuccinimide Ester) 50 mg 416.30 Maleimide, Sulfo-NHS Ester 9.9 Å Amines, Sulfhydryls 22312 PI22312 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Sulfo-SANPAH (sulfosuccinimidyl 6-(4'-azido-2'-nitrophenylamino)Hexanoate) 50 mg 492.40 Nitrophenyl azide and Sulfo-NHS 18.2 Å Amines, Sulfhydryls 22589 PI22589 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Sulfo-SIAB (Sulfosuccinimidyl-(4-iodoacetyl)Aminobenzoate) 50 mg 504.19 Iodoacetyl, Sulfo-NHS Ester 10.6 Å Amines, Sulfhydryls 22327 PI22327 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Sulfo-SMCC (Sulfosuccinimidyl-4(N-maleimidomethyl) Cyclohexane-1-carboxylate) 50 mg 436.37 Maleimide, Sulfo-NHS Ester 8.3 Å Amines, Sulfhydryls 22322 PI22322 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Sulfo-SMPB (Sulfosuccinimidy-4-(p-maleimidophenyl)Butyrate) 50 mg 458.38 Maleimide, Sulfo-NHS Ester 14.5 Å Amines, Sulfhydryls 22317 PI22317 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
View MSDS SDS
Sulpho-SMCC (Sulphosuccinimidyl 4-(N-maleimidomethyl)Cyclohexane-1-carboxylate) 1 g 436.37 Maleimide, Sulfo-NHS Ester 8.3 Å Amines, Sulfhydryls 22122 PI22122 Each (1g) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
TFCS (N-(Ɛ-Trifluoroacetylcaproyloxy)Succinimide Ester) 50 mg 324.25 NHS Ester, Trifluoroacetyl-Protected -NH2 7.7 Å Amines, NHS/Sulfo-NHS Esters, EDC-Activated Carboxyls 22299 PI22299 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Tris-(2-Maleimidoethyl)amine (Trifunctional) 50 mg 386.36 Maleimide 10.3 Å Sulfhydryls 33043 PI33043 Each (50mg) Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
Tris-(succimimidyl aminotricetate) (Trifunctional) 50 mg 482.36 NHS Ester 4.2 Å Amines 33063 PI33063 Each Retrieving
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty