Erk inhibitor ≥95% (by HPLC), Calbiochem®

Supplier: MilliporeSigma

328006-5MG 328006-5MG
80016-986EA 148.84 USD
80016-986 CA80016-986
Erk inhibitor ≥95% (by HPLC), Calbiochem®
Erk inhibitor

3-(2-Aminoethyl)-5-((4-ethoxyphenyl)methylene)-2,4-thiazolidinedione, HCl


  • DMSO (10 mg/mL)


Light yellow solid. A cell-permeable thiazolidinedione compound that displays anti-proliferative properties. It preferentially binds to ERK2 with a KD of ~5µM and prevents its interaction with protein substrates. It is also shown to block ERK-mediated phosphorylation of ribosomal S6 kinase-1 (RSK-1) and ternary complex factor ELK-1 in HeLa cells, while exhibiting little effect on ERK1/2 phosphorylation by MEK1/2.


Packaged under inert gas. Protect from light. Hygroscopic. Store at 4°C (39°F). Following reconstitution, aliquot and store at –20°C (–4°C). Stock solutions are stable for up to three months when frozen. For research use only, not for therapeutic or diagnostic use.

Formula: C₁₄H₁₆N₂O₃S·HCl
MW: 328.8 g/mol
CAS Number: 1049738-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR