eLINE® Multichannel Electronic Pipettors, Variable Volume, Sartorius
Supplier: Sartorius
eLINE®
730391 730361 730441
CA89171-658 CA89171-654 CA89171-652
eLINE® Multichannel Electronic Pipettors, Variable Volume, Sartorius
Pipettes Micropipettes
Sartorius eLINE® Electronic Pipettors are made to deliver years of real performance and total user convenience.
  • Proven accuracy and precision with DC-motor concept
  • Fully electronic operations guarantee user-independent results and high ergonomics to prevent RSI (Repetitive Strain Injury)
  • Comprehensive range of pipetting modes
  • Universal AC adaptor included (EU, UK, US/JPN, and CHN plugs)
They feature a comprehensive range of pipetting modes and are twice as fast as mechanical pipettors. One eLINE® pipettor can be used for several liquid handling tasks, from regular pipetting to diluting with mixing and automatic repetitive dispensing. The comprehensive range of pipetting modes in eLINE® reduces the need for several work stages and enables liquid dispensing significantly faster than with a mechanical pipettor. The DC-motor concept, with built-in error control, improves pipetting precision and enables reliable results.