You Searched For: Crystal Violet


38  results were found

SearchResultCount:"38"

Sort Results

List View Easy View

Rate These Search Results

Supplier: MilliporeSigma
Description: HARLECO® Individual replacement for crystal violet component of the Gram Staining Set. Stains Gram-positive bacteria purple
Supplier: MilliporeSigma
Description: CRYSTAL VIOLET (C.I. 425 (25G)
Catalog Number: (34172-132)
Supplier: MilliporeSigma
Description: Used for staining bacteria and spores by Gram’s method.

Supplier: Ricca Chemical
Description: CRYSTAL (GENTIAN) VIOLET 1% IN
Supplier: TCI America
Description: CAS Number: 548-62-9 MDL Number: MFCD00011750 Molecular Formula: C25H30ClN3 Molecular Weight: 407.99 Form: Crystal Color: Yellow Green Melting poi...
Supplier: Ricca Chemical
Description: GENTIAN VIOLET 1% (W/V) ALCOH
Catalog Number: (RC2480-4)
Supplier: Ricca Chemical
Description: CRYSTL VIOLET 0.2%IN GAA 4OZ

Supplier: Ricca Chemical
Description: Gentian Violet, Hucker Formulation, for Gram Staining
Supplier: Ricca Chemical
Description: Crystal Violet-Acetic Acid Solution for Cerebrospinal Fluid Cell Count
Catalog Number: (89405-046)
Supplier: Hardy Diagnostics
Description: Used as a primary stain in the advanced gram stain procedure

Product available on GSA Advantage®


Catalog Number: (TCC2460-5G)
Supplier: TCI America
Description: [for Biochemical Research] CAS Number: 548-62-9 MDL Number: MFCD00011750 Molecular Formula: C25H30ClN3 Molecular Weight: 407.99 Form: Crystal Colo...

Supplier: Ricca Chemical
Description: GENTIAN VIOLET 2% (W/V) ALCOH
Catalog Number: (RC2479-4)
Supplier: Ricca Chemical
Description: CRYSTAL VIOLET 0.1% IN GAA 4OZ

Supplier: Azer Scientific Inc.
Description: Primary stain in the Gram Stain method for differentiating bacteria.

Product available on GSA Advantage® Minority or Woman-Owned Business Enterprise

Catalog Number: (76459-552)
Supplier: bioMérieux
Description: Color Gram 2, R1 Oxalate Crystal Violet, used to stain bacterial and fungal microorganisms for direct examination, according to the Gram method. For manual Gram Staining, 1 x 2 Litre


Supplier: Thermo Scientific Chemicals
Description: 500g.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 38
no targeter for Bottom