VWR® 96-Well Thermal Cycling Plates
Supplier: VWR International
VWR®
3450-00-521AV 3450T00-521AV
89049-178CS 398.29 USD InStock
89049-178 89218-290
VWR® 96-Well Thermal Cycling Plates
Microplates PCR Plates
Plates feature a smooth rim that allows better contact inside the block to ensure a perfect fit and easy removal, as well as printed alphanumeric labeling for easy well location.
  • Inert polypropylene construction
  • Each well holds 200 µL
Designed to fit ABI PRISM® 3700 series thermocyclers.