Combination Platinum Oxidation Reduction Potential Electrode, Gel-Filled, Beckman Coulter®
Supplier: Beckman Coulter
A57197 A57197
BKA57197EA 404.21 USD
BKA57197 CABKA57197
Combination Platinum Oxidation Reduction Potential Electrode, Gel-Filled, Beckman Coulter®
Electrodes Redox Electrodes
This electrode is for oxidation reduction potential (ORP) measurements.
  • One-Meter Cable with BNC Connector
  • Protective Epoxy Body
  • For oxidation reduction potential (ORP) measurements
A rugged epoxy body extends over glass bulb for protection.
The electrode has an attached, 1 m cable with a BNC connector.