Petri Dish Dispensers and Racks, Mitchell Plastics™
Supplier: Mitchell Plastics
ML-3500 ML-3000-1 ML-3000W,44 ML-3000W44-1 ML-3200-1 ML-3050-1 ML-3000
89094-528EA 421.9 USD
89094-528 25405-203 25405-414 25405-206 89094-526 89094-538 89094-532
Petri Dish Dispensers and Racks, Mitchell Plastics™
Racks Petri Dish Racks
For stacking, storing, and transporting empty and prepared petri dishes
Wall-mountable dispensers feature a dispensing slot at the base for easy removal of dishes. Benchtop racks feature a carrying handle. Dispensers and racks are made from clear 6mm (1/4") thick acrylic. Not autoclavable.