Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PCR Tubes and Tube Strips, Greiner Bio-One

Supplier: GREINER BIO-ONE
673277 682273 682274 682272 682201 684201 683276 683277 682281 683271 683274 683275 683272 683273 682277 683201 682275 682276 673273 673274 673275 673210 673276 673271 673272
Polypropylene tubes feature thin-wall construction for optimal heat transfer from the thermocycler block to the reaction mixture.

  • Polypropylene construction
  • Thin-walled construction
  • Nonpyrogenic
  • Free of RNase and DNase

Tight-fitting caps, available flat or domed, ensure low evaporation.

Tubes are nonpyrogenic and RNase- and DNase-free.

Ordering information: Individual tubes feature attached caps, except for 82051-064, which has no cap. Assorted packs consist of blue, green, orange, red, violet, and yellow. The eight-tube strips are supplied without caps. Eight strip tube caps are available separately (see 89233-706 series).




 

ORDER

Expand All / Collapse All

Tubes
Description Format Volume Cap Type Color Supplier No. VWR Catalog Number Unit Price Quantity
Individual Tube Tubes 0.5 mL Flat Clear 682201 82051-032 Case of 1,000 Retrieving
Individual Tube Tubes 0.2 mL Flat Clear 683201 82051-048 Case of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty