Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PCR Clean Microcentrifuge Tubes, Eppendorf®

Supplier: Eppendorf
Flex-Tube® Safe-Lock
022363719 022364120 022363344 022363212
Microcentrifuge tubes are constructed of highest purity, low adhesion polypropylene

The easy-to-open, securely closing, Eppendorf® Flex-Tube® offers comfort and dependability for sample preperation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry.

The original Salfe-Lock Tubes protect samples with excellent centrifugation stability, feature perfect dealing properties, and reliably prevent sample evaporation.

With a flat top, volume graduations, and etched marking surfaces, tubes feature secure sealing, even while boiling. These Flex-Tubes® and Safe-Lock tubes meet Eppendorf® PCR clean standards; free of human DNA, DNase, RNase, and PCR inhibitors for all molecular biology applications. Production and packaging is fully automated in a class 8 cleanroom environment. Batches are tested and certified by an independent laboratory.




 

ORDER

Expand All / Collapse All
Description Volume Maximum RCF Supplier No. VWR Catalog Number Unit Price Quantity
Flex-Tube® 1.5 mL 30000 ×g 022364120 62111-756 Pack of 500 Retrieving
Product available on GSA Advantage®
Safe-Lock Tube 0.5 mL 30000 ×g 022363719 62111-750 Pack of 500 Retrieving
Product available on GSA Advantage®
Safe-Lock Tube 1.5 mL 30000 ×g 022363212 62111-752 Pack of 500 Retrieving
Product available on GSA Advantage®
Safe-Lock Tube 2.0 mL 25000 ×g 022363344 62111-754 Pack of 500 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty