Would you like to log in again?
PCR Tubes with PurePak™ Packaging, Molecular BioProducts
Supplier: Thermo Scientific
PurePak™
3412Y 3414 3412 3430 3414B 3412P 3418B 3418
Quality-inspected, thin-walled tubes are certified RNase-, DNase-, DNA-, and pyrogen-free
Tubes are compatible with most major brands of thermal cyclers.
Made of premium-grade polypropylene, tubes feature tightly sealing caps to minimize sample evaporation during thermal cycling. Thin-wall construction ensures uniform contact with the thermal cycler block, producing optimal heat transfer. This reduces cycle times and helps maintain accurate reaction temperatures. PurePak™ packaging keeps PCR tubes contamination-free. Each pack is divided into individually sealed bags, ensuring the purity of unused tubes.
ORDER
Expand All / Collapse All
Description Format Volume Cap Type Color Supplier No. VWR Catalog Number Unit Price Quantity
Individual PCR Tube with Dome Cap Tubes 0.2 mL Domed Clear 3414 53225-640 Case of 10000 Retrieving
Pack of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty