Filling Solutions, Beckman Coulter®
Supplier: Beckman Coulter
511178 511179 511177
BK511177EA 47.25 USD
CABK511178 CABK511179 BK511177
Filling Solutions, Beckman Coulter®
Electrolytes Electrode Filling Solutions
Filling solutions for the various ions are provided in 125 mL bottles.
  • Application: Determining Specific Ions
  • Refillable Epoxy Body Construction
  • Solutions in 125 mL Bottles
  • Integrated Cable
  • BNC Connector
Filling solutions determine specific ions.
All electrodes feature refillable epoxy body construction and an
integrated, one-meter cable with a BNC connector.