Cytochalasin B (from Helminthosporium dematioideum) ≥98% (by HPLC), Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 7(S),20(R)-Dihydroxy-16(R)-methyl-10-phenyl-24-oxa[14]cytochalasa-6(12),13(E),21(E)-triene-1,23-dione

250233-5MG 250233-5MG
80055-212EA 233.73 USD
80055-212 CA80055-212
Cytochalasin B (from Helminthosporium dematioideum) ≥98% (by HPLC), Sigma-Aldrich®
Cytochalasin B

Cell-permeable fungal toxin


Inhibits cytoplasmic division by blocking the formation of contractile microfilaments. Inhibits cell movement and induces nuclear extrusion. Shortens actin filaments by blocking monomer addition at the fast-growing end of polymers. Inhibits glucose transport and platelet aggregation. Blocks adenosine-induced apoptotic body formation without affecting activation of endogenous ADP-ribosylation in leukemia HL-60 cells.


Soluble in MeOH or DMSO

Formula: C₂₉H₃₇NO₅
MDL Number: MFCD00077704
CAS Number: 14930-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR