VWR® SuperClear™ Ultra-High Performance Freestanding Centrifuge Tubes with Flat Caps, Polypropylene
3095-870-008
21008-480CS 176.44 USD
21008-480
VWR® SuperClear™ Ultra-High Performance Freestanding Centrifuge Tubes with Flat Caps, Polypropylene
These disposable polypropylene tubes have smooth inner walls with external printed (stamped) graduations for minimal sample hang-up.
  • RNase-, DNase-, endotoxin-free
  • Autoclavable
  • Freezable
Validated ISO 11137 Sterile Process.
Ordering information: Supplied with polyethylene flat caps. Caps are not autoclavable.