Tunicamycin (from Streptomyces lysosuperficus), Calbiochem®

Supplier: MilliporeSigma

654380-50MG 654380-10MG 654380-10MG 654380-50MG
80055-486EA 854.93 USD
CA80055-484 80055-486 CA80055-486 80055-484
Tunicamycin (from Streptomyces lysosuperficus), Calbiochem®
Tunicamycin

A nucleoside antibiotic that inhibits N-linked glycosylation and blocks the formation of N-glycosidic protein-carbohydrate linkages


Active in vitro against gram-positive bacteria, yeasts, fungi, and viruses. Inhibits the expression of functional thrombin receptors on human T-lymphoblastoid cells. Also inhibits thrombin-induced Ca2+ mobilization in cells. Aqueous solutions can be prepared from stock solutions in DMSO by dilution with water at pH 8.0 to 10.0 or with appropriate buffers at pH >7.0 (preferably pH >8.0).

Formula: C₃₉H₆₄N₄O₁₆
MW: 844.95 g/mol
MDL Number: MFCD00065709
CAS Number: 11089-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR