Streptozocin, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Streptozotocin

572201-1GM 572201-250MG
80054-530EA 132.79 USD
80054-530 80054-528
Streptozocin, Calbiochem®
Streptozocin

An N-nitroso-containing diabetogenic agent compound that acts as a nitric oxide donor in pancreatic islets


An antibiotic effective against gram-negative bacteria. Causes DNA alkylation and DNA strand breaks in pancreatic islet cells.

Formula: C₈H₁₅N₃O₇
MW: 265.2 g/mol
MDL Number: MFCD00006607
CAS Number: 18883-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR