Monensin monosodium salt hydrate, high purity, Calbiochem®
475895-250MG
80055-340EA 255.13 USD
80055-340
Monensin monosodium salt hydrate, high purity, Calbiochem®
Monensin monosodium salt hydrate
Polyether antibiotic that functions as a Na+ ionophore by forming stable complexes with monovalent cations that are able to cross the plasma membrane
Blocks glycoprotein secretion. Reduces sphingomyelinase activity. Inhibits the ability of HDL to decrease cellular cholesterol esterification. Also reported to induce catecholamine secretion from adrenal chromaffin cells.