Mevastatin ≥95% (by HPLC), Calbiochem®
Supplier: MilliporeSigma
Synonyms:
(1S,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (S)-2-methylbutanoate
, Compactin
474700-50MG
474700-50MG
80057-044EA
176.46
USD
80057-044
CA80057-044
Mevastatin ≥95% (by HPLC), Calbiochem®
Mevastatin
An antibiotic that acts as a potent inhibitor of HMG-CoA reductase, thus suppressing Ras farnesylation
- White solid
Inhibitor of myoblast fusion. Causes cell cycle arrest in late G1 phase. This is an inactive form. To activate, treat with NaOH in ethanol and then neutralize to pH 7.2.
Soluble in EtOH
Formula:
C₂₃H₃₄O₅ MW: 390.52 g/mol |
MDL Number:
MFCD05662341 CAS Number: 73573-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...