Tetracycline, Sigma-Aldrich®

58346-25GM 58346-10GM 58346-10GM 58346-50GM 58346-25GM 58346-50GM
80058-724EA 99.4 USD
80058-724 CA80108-314 80108-312 CA80058-724 CA80108-312 80108-314
Tetracycline, Sigma-Aldrich®
Tetracycline

Broad-based antibiotic agent that blocks protein synthesis by inhibiting binding of aminoacyl tRNA to the A-site of ribosomes


Induces cold shock-response and enhances P450 expression in bacteria. Activity: >=900µg/mg.

Formula: C₂₂H₂₄N₂O₈
MDL Number: MFCD00151232
CAS Number: 60-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR