Carbenicillin disodium salt, Calbiochem®, Millipore®

Synonyms: 6-[(Carboxyphenylacetyl)-amino]-3,3-dimethyl-7-oxo-4-thia-1-aza-bicyclo[3.2.0]heptane-2-carboxylic Acid, 2Naα-Carboxybenzylpenicillin disodium salt

M-Clarity™ quality level = MQ100

205805-250MG 205805-250MG
80057-880EA 75.91 USD
80057-880 CA80057-880
Carbenicillin disodium salt, Calbiochem®, Millipore®
Carbenicillin disodium salt

Synthetic penicillin derivative for ampicillin-resistant plasmids. Blocks bacterial cell wall formation. Effective against Gram-negative bacteria.

Formula: C₁₇H₁₆N₂O₆S·2Na
MW: 422.363 g/mol
Storage Temperature: Ambient
CAS Number: 4800-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR